Z663670
Brady® thermal labeling system (TLS) labels
general industrial, W × L 25.40 mm × 15 m, yellow vinyl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
yellow vinyl
packaging
roll of 15 m
manufacturer/tradename
Brady PTL-42-439-YL
availability
available only in EU
W × L
25.40 mm × 15 m
Looking for similar products? Visit Product Comparison Guide
Legal Information
Brady is a registered trademark of Brady Worldwide, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service