Z353353
Freezing vials, sterile
capacity 2.0 mL, skirted bottom
Synonym(s):
cryopreservation tubes, cryopreservation vials, cryovials, freezing tubes
About This Item
Recommended Products
material
colorless tube
skirted bottom
description
extrenal threads
sterility
sterile
sterile
feature
External thread
packaging
pack of 500 ea
parameter
-196 °C min. temp.
technique(s)
cryopreservation: suitable
capacity
2.0 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Caps are supplied with attachment loops, and allow them to remain attached to the tube in order to prevent mix-up and contamination.
- Perfect for cryogenic work
- Non skirted tubes can be centrifuged up to 20,000g. (skirted 17,000g.)
- Will withstand temperatures from -196°C to +121°C
Please note: Do not immerse during liquid phase of liquid nitrogen.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service