Skip to Content
Merck
All Photos(1)

Key Documents

TMO374025

Nunc® Coded Cryobank Vials in black low rack

polypropylene, capacity 0.5 mL, self-standing natural, sterile, blue cap, case of 960 ea

Synonym(s):

coded cryobank vial storage, cryostorage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121806
NACRES:
NB.11

material

blue cap
polypropylene
self-standing natural

sterility

sterile

packaging

case of 960 ea

manufacturer/tradename

Nunc 374025

technique(s)

cryopreservation: suitable

capacity

0.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Related applications: Cell Culture Plastics

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service