SRP5210
FLT4 (800-end), active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLT41, LMPH1A, PCL, VEGFR3
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
57.8-78.2 nmol/min·mg
mol wt
~85 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... FLT4(2324)
General description
FLT4 or fms-related tyrosine kinase 4 is a tyrosine kinase receptor for vascular endothelial growth factors C and D and highly expressed in lymphangiogenesis and maintenance of the lymphatic endothelium. FLT4 is a marker for lymphatic vessels and some high endothelial venules in human adult tissues and also supported the theory of the venous origin of lymphatic vessels. FLT4 plays an essential role in the development of the embryonic cardiovascular system before the emergence of the lymphatic vessels.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Kaipainen et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(8), 3566-3570 (1995-04-11)
We have recently cloned the human fms-like tyrosine kinase 4 gene FLT4, whose protein product is related to two vascular endothelial growth factor receptors FLT1 and KDR/FLK1. Here the expression of FLT4 has been analyzed by in situ hybridization during
D J Dumont et al.
Science (New York, N.Y.), 282(5390), 946-949 (1998-10-30)
Vascular endothelial growth factor (VEGF) is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGF-C stimulates the growth of lymphatic vessels through its specific lymphatic endothelial receptor VEGFR-3. Here it
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service