Skip to Content
Merck
All Photos(1)

Key Documents

SRP0683

Sigma-Aldrich

RS Repeat Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

product name

RS Repeat Peptide, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized powder

mol wt

1934.13

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

The synthetic peptide (GRSRSRSRSRSRSRSR) contains serine/threonine protein kinase phosphorylation sites and is routinely evaluated as a substrate for CLK, EIF2AK and SRPK family kinases.

Sequence

GRSRSRSRSRSRSRSR

Reconstitution

Reconstitute in 25 mM Tris-HCl, pH 7.5 solution to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at --20°C. Avoid freeze/thaw cycles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service