Skip to Content
Merck
All Photos(1)

Key Documents

SML3609

Sigma-Aldrich

Sulthiame

≥95% (HPLC)

Synonym(s):

4-(1,1-Dioxothiazinan-2-yl)benzenesulfonamide, 4-(Tetrahydro-1,1-dioxido-2H-1,2-thiazin-2-yl)-benzenesulfonamide, 4-sulfamoyl-phenyl-[1,2]thiazinane-1,1-dioxide, Sultiame

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O4S2
CAS Number:
Molecular Weight:
290.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

NS(C1=CC=C(N2CCCCS2(=O)=O)C=C1)(=O)=O

InChI

1S/C10H14N2O4S2/c11-18(15,16)10-5-3-9(4-6-10)12-7-1-2-8-17(12,13)14/h3-6H,1-2,7-8H2,(H2,11,15,16)

InChI key

HMHVCUVYZFYAJI-UHFFFAOYSA-N

Biochem/physiol Actions

Carbonic anhydrase inhibitor with in vivo anticonvulsant (antiepileptic) efficacy.
Sulthiame is a carbonic anhydrase (CA) inhibitor (hCA Ki in nM = 374/I, 7/II, 95/IV, 81/VA, 91/VB, 134/VI, 6/VII, 43/IX, 56/XII; Ki = 630 mM/hCA III, 1.45 μM/mCA XIII, 1.54 μM/hCA XIV) with anticonvulsant efficacy in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the Sulfonamide Inhibition Profiles of the ?- and ?-Carbonic Anhydrases from the Pathogenic Bacterium Burkholderia pseudomallei
Molecules (Basel), 22(3), 421-421 (2017)
Claudia Temperini et al.
Bioorganic & medicinal chemistry letters, 17(17), 4866-4872 (2007-06-26)
Sulthiame, a clinically used antiepileptic, was investigated for its interaction with 12 catalytically active mammalian carbonic anhydrase (CA, EC 4.2.1.1) isoforms. The drug is a potent inhibitor of CA II, VII, IX, and XII (K(I)s of 6-56 nM), and a
Tobias Leniger et al.
Epilepsia, 43(5), 469-474 (2002-05-25)
Sulthiame is a carbonic anhydrase (CA) inhibitor with an anticonvulsant effect in the treatment of benign and symptomatic focal epilepsy in children. The aim of the study was to elucidate the mode of action of sulthiame with respect to possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service