Skip to Content
Merck
All Photos(1)

Key Documents

SML2825

Sigma-Aldrich

I-BET469

≥98% (HPLC)

Synonym(s):

5-(1-(1,3-Dimethoxypropan-2-yl)-5-morpholino-1H-benzo[d]imidazol-2-yl)-1,3-dimethylpyridin-2(1H)-one, 5-[1-[2-Methoxy-1-(methoxymethyl)ethyl]-5-(4-morpholinyl)-1H-benzimidazol-2-yl]-1,3-dimethyl-2(1H)-pyridinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N4O4
CAS Number:
Molecular Weight:
426.51
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N4(CCOCC4)c1cc2nc([n](c2cc1)C(COC)COC)C3=CN(C(=O)C(=C3)C)C

InChI

1S/C23H30N4O4/c1-16-11-17(13-25(2)23(16)28)22-24-20-12-18(26-7-9-31-10-8-26)5-6-21(20)27(22)19(14-29-3)15-30-4/h5-6,11-13,19H,7-10,14-15H2,1-4H3

InChI key

DDSHVVXWYKDTTQ-UHFFFAOYSA-N

Biochem/physiol Actions

I-BET469 is an orally active bromodomain (BrD) inhibitor with selectivity toward a subset of bromodomain-containng proteins (BCPs) and BrDs (Kd in nM: BRD4 BrD1/2 = 1.5/2.9, BRD3 BrD1/2 = 2/2.3, BRD2 BrD1/2 = 2.3/2.8, BRDT BrD1/2 = 12/100, CREBBP = 37, EP300 = 49; Kd ≥4.8 μM toward 22 other BCPs). I-BET469 targets cellular BRD4 with high affinity (pKd = 8.3 or Kd = 5.0 nM by NanoBRET) and shows in vivo efficacy in a murine acute endotoxic shock model (1, 3, 10 mg/kg po. 30 min prior to LPS) and a chronic T cell-dependent immunization model (IgG suppression on day 14 = 24% and 65%, respectively, with 0.3 mg/kg or 3 mg/kg daily po.).
Orally active bromodomain (BrD) inhibitor with in vivo efficacy and selectivity for BRD2/3/4, BRDT, CREBBP, EP300 over 22 other BCPs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher R Wellaway et al.
Journal of medicinal chemistry, 63(2), 714-746 (2020-01-07)
The bromodomain and extraterminal (BET) family of bromodomain-containing proteins are important regulators of the epigenome through their ability to recognize N-acetyl lysine (KAc) post-translational modifications on histone tails. These interactions have been implicated in various disease states and, consequently, disruption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service