Skip to Content
Merck
All Photos(1)

Key Documents

SML1423

Sigma-Aldrich

Pranoprofen

≥98% (HPLC)

Synonym(s):

2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid, 2-(5H-[1]-Benzopyrano[2,3-b]pyridin-7-yl)propionic acid, a-Methyl-5H-[1]benzopyrano[2,3-b]pyridine-7-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3
CAS Number:
Molecular Weight:
255.27
MDL number:
UNSPSC Code:
41106500
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C(C)C1=CC2=C(C=C1)OC3=NC=CC=C3C2)=O

InChI

1S/C15H13NO3/c1-9(15(17)18)10-4-5-13-12(7-10)8-11-3-2-6-16-14(11)19-13/h2-7,9H,8H2,1H3,(H,17,18)

InChI key

TVQZAMVBTVNYLA-UHFFFAOYSA-N

General description

Pranoprofen is an arylalkanoic acid and a non-steroidal anti-inflammatory drug.

Biochem/physiol Actions

Pranoprofen is a non-steroidal anti-inflammatory drug (NSAID) used primarily in ophthalmology. Pranoprofen is a cyclooxygenase (COX) inhibitor.
Pranoprofen is a non-steroidal anti-inflammatory drug (NSAID).
Pranoprofen is used in eyes to treat chronic allergic conjunctivitis. It is also used as an anti-inflammatory and analgesic drug after strabismus surgery.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service