Skip to Content
Merck
All Photos(1)

Key Documents

SML1379

Sigma-Aldrich

Seratrodast

≥98% (HPLC)

Synonym(s):

(±)-7-(3,5,6-Trimethyl-1,4-benzoquinon-2-yl)-7-phenylheptanoic acid, AA-2414

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O4
CAS Number:
Molecular Weight:
354.44
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(C)=C1C)C(C)=C(C(CCCCCC(O)=O)C2=CC=CC=C2)C1=O

InChI

1S/C22H26O4/c1-14-15(2)22(26)20(16(3)21(14)25)18(17-10-6-4-7-11-17)12-8-5-9-13-19(23)24/h4,6-7,10-11,18H,5,8-9,12-13H2,1-3H3,(H,23,24)

InChI key

ZBVKEHDGYSLCCC-UHFFFAOYSA-N

Gene Information

human ... TBXA2R(6915)

Biochem/physiol Actions

Seratrodast is a thromboxane A2 (TXA2) receptor (TP receptor) antagonist with anti-asthmatic activity.
Seratrodast is a thromboxane A2 (TXA2) receptor (TP receptor) antagonist with anti-asthmatic activity. Seratrodast inhibits the bronchconstriction induced by leukotriene D4 (LTD4), and platelet activating factor (PAF) with a long duration of action.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarita Rosa Romero-Imbachi et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 240, 108939-108939 (2020-11-10)
Centruroides margaritatus scorpion stings are common in Colombia. However, the cardiovascular toxicity of the venom has not been clarified. To study the effect and mechanisms of action of the complete venom of C. margaritatus (CmV) on the murine cardiovascular system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service