Skip to Content
Merck
All Photos(1)

Key Documents

SML1101

Sigma-Aldrich

Nedocromil sodium

≥98% (HPLC)

Synonym(s):

9-Ethyl-6,9-dihydro-4,6-dioxo-10-propyl-4H-pyrano[3,2-g]quinoline-2,8-dicarboxylic acid sodium salt, Nedocromil disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO7 · 2 Na
CAS Number:
Molecular Weight:
415.30
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, white to beige to brown

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[n]1(c2c([c](cc1C(=O)[O-])=O)cc3c([o]c(c[c]3=O)C(=O)[O-])c2CCC)CC

InChI

1S/C19H17NO7.2Na/c1-3-5-9-16-10(13(21)7-12(18(23)24)20(16)4-2)6-11-14(22)8-15(19(25)26)27-17(9)11;;/h6-8H,3-5H2,1-2H3,(H,23,24)(H,25,26);;/q;2*+1/p-2

InChI key

JQEKDNLKIVGXAU-UHFFFAOYSA-L

Biochem/physiol Actions

Nedocromil is a mast cell stabilizer and anti-inflammatory and anti-allergic drug.
Nedocromil is a mast cell stabilizer and anti-inflammatory and anti-allergic drug. Nedocromil inhibits the degranulation of mast cells, and prevents the activation of and mediator release from a variety of inflammatory cell types. Nedocromil inhibits release of histamine, leukotriene C4, and prostaglandin D2, but has no antihistamine, bronchodilator, or corticosteroid activity itself.
Nedocromil is considered to be more effective in treating asthma compared to a placebo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is nedocromil sodium effective treatment for asthma?
O'Byrne P M and Cook D
The European Respiratory Journal, 6(1), 5-6 (1993)
Ajantha Sinniah et al.
International immunopharmacology, 32, 87-95 (2016-01-25)
1.We investigated the role of Annexin (ANX)-A1 and its receptor, ALX/FPR2, in the regulation of mast cell degranulation produced by compound 48/80. 2.Both human cord-blood derived mast cells (CBDMCs) and murine bone marrow derived mast cells (BMDMCs) release phosphorylated ANX-A1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service