Skip to Content
Merck
All Photos(1)

Documents

SMB00323

Sigma-Aldrich

Schisandrin C

≥98% (HPLC)

Synonym(s):

Schisandrin C, Schizandrin C, Wuweizisu C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24O6
CAS Number:
Molecular Weight:
384.42
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COC1=C2C(OCO2)=CC3=C1C(C(OC)=C(OCO4)C4=C5)=C5CC(C)C(C)C3

InChI

1S/C22H24O6/c1-11-5-13-7-15-19(27-9-25-15)21(23-3)17(13)18-14(6-12(11)2)8-16-20(22(18)24-4)28-10-26-16/h7-8,11-12H,5-6,9-10H2,1-4H3

InChI key

HTBWBWWADZJXID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Schisandrin C is a lignan found in the fruit of Schisandra chinensis.

Biochem/physiol Actions

Schisandrin C has anti-inflammatory effects and has been shown to reduce nitric oxide (NO) production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jibo Han et al.
Phytotherapy research : PTR, 33(3), 779-790 (2019-01-12)
Vascular endothelial dysfunction plays a crucial role in the pathogenesis of cardiovascular diseases. Oxidative stress is a key pathophysiological mechanism underpinning endothelial dysfunction. Schisandrin C (Sch C), a dibenzocyclooctadiene derivative of Schisandra chinensis, has antioxidative properties. Here, we report the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service