All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H41N3O5
Molecular Weight:
475.62
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥85%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Z-Leu-Leu-Nle-al
General description
Gamma-secretase is a protein complex containing presenilin, nicastrin, APH-1, and PEN-2. The leupeptin analogue Z-Leu-Leu-Nle-al (N-benzyloxycarbonyl-Leu-Leu-norleucinal; Benzyloxicarbonyl-Leu-Leu-Nle-CHO (LLNle)) and closely related peptides are used as protease and a gamma secretase inhibitors.
Application
γ Secretase Inhibitor has been used to study the suppressive miR-129 functions through inhibition on estrogen receptor 1 (ESR1) and NOTCH signaling in breast cancer stem cells.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cleonice Michelon et al.
Molecular neurobiology, 57(2), 1159-1169 (2019-11-09)
Inflammatory cytokines are related to impaired learning and memory processes in the central nervous system, contributing to the cognitive dysfunction present in sepsis survivors. In sepsis, brain of survivors presented increased deposition of amyloid-beta (Aβ) peptide and this was associated
MiR-129 blocks estrogen induction of NOTCH signaling activity in breast cancer stem-like cells.
Xiao G, et al.
Oncotarget, 8(61), 103261-103261 (2017)
Tao Na et al.
BMC cancer, 20(1), 1064-1064 (2020-11-06)
Immunomodulatory activities of human mesenchymal stromal /stem cells (hMSCs) has been widely recognized as the most critical function of hMSCs for exerting its therapeutic effects. However, the detailed mechanisms responsible for regulating the immunomodulation of hMSCs still remain largely unknown.
Asha Acharya et al.
PloS one, 6(11), e27743-e27743 (2011-11-24)
Aortic valve calcification is the most common form of valvular heart disease, but the mechanisms of calcific aortic valve disease (CAVD) are unknown. NOTCH1 mutations are associated with aortic valve malformations and adult-onset calcification in families with inherited disease. The
Guodong Xiao et al.
Oncotarget, 8(61), 103261-103273 (2017-12-22)
Stem-like cells in tumor group featured the major role in the chemotherapy resistance of breast cancer, and the reduction of stem-like cells helped to perish the tumor when receiving chemotherapy. Smaller stem cells number indicated better therapeutic effect in vitro
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service