Skip to Content
Merck
All Photos(2)

Documents

SAB4503292

Sigma-Aldrich

Anti-CSPN3 (Cleaved-Asp175) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Apopain precursor, CASP-3, CPP-32, CPP32, Caspase-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 20 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Asp175)

Gene Information

human ... CASP3(836)

General description

Anti-ANTI-CSPN3 (Cleaved-Asp175) Antibody detects endogenous levels of fragment of activated ANTI-CSPN3 (Cleaved-Asp175) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Caspase 3.

Immunogen Range: 126-175

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhu et al.
FEBS open bio, 13(10), 1859-1873 (2023-08-01)
Sepsis-induced acute kidney injury (SI-AKI) causes renal dysfunction and has a high mortality rate. Protein arginine methyltransferase-1 (PRMT1) is a key regulator of renal insufficiency. In the present study, we explored the potential involvement of PRMT1 in SI-AKI. A murine
Ming Tu et al.
Molecular medicine reports, 15(4), 1713-1721 (2017-03-06)
Cluster of differentiation 164 (CD164), a sialomucin, has been demonstrated to be involved in the regulation of proliferation, apoptosis, adhesion and differentiation in multiple cancers. CD164 is regarded to be a potential promotor of tumor growth. However, the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service