Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500321

Sigma-Aldrich

Anti-DNMT1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CXXC9, Anti-DNA (cytosine-5-)-methyltransferase 1, Anti-DNMT, Anti-FLJ16293, Anti-MCMT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, mouse, bovine, canine, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... DNMT1(1786)

Related Categories

Immunogen

Peptide with sequence C-RFESPPKTQPTEDN from the internal egion of the protein sequence according to NP_001370.1 .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Li et al.
Chinese journal of cancer research = Chung-kuo yen cheng yen chiu, 26(4), 371-381 (2014-09-19)
To better understand the contribution of dysregulated DNA methyltransferase 1 (DNMT1) expression to the progression and biology of clear cell renal cell carcinoma (ccRCC). We examined the differences in the expression of DNMT1 in 89 ccRCC and 22 normal tissue
Mina Sasaki et al.
Biochemical and biophysical research communications, 452(3), 622-628 (2014-09-03)
Reactive oxygen species (ROS) can cause severe damage to DNA, proteins and lipids in normal cells, contributing to carcinogenesis and various pathological conditions. While cellular senescence arrests the early phase of cell cycle without any detectable telomere loss or dysfunction.
Vanesa G Martinez et al.
Cellular & molecular immunology, 11(4), 343-354 (2014-03-04)
Apoptosis inhibitor of macrophages (AIMs), a homologue of human Spα, is a mouse soluble member of the scavenger receptor cysteine-rich superfamily (SRCR-SF). This family integrates a group of proteins expressed by innate and adaptive immune cells for which no unifying
Bo Ra You et al.
Molecular carcinogenesis, 53(11), 847-857 (2013-05-11)
Zebularine (Zeb) is a DNA methyltransferase (DNMT) inhibitor to that has an anti-tumor effect. Here, we evaluated the anti-growth effect of Zeb on A549 lung cancer cells in relation to reactive oxygen species (ROS) levels. Zeb inhibited the growth of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service