Skip to Content
Merck
All Photos(2)

Key Documents

SAB2107930

Sigma-Aldrich

Anti-SCN5A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CDCD2, Anti-CMD1E, Anti-CMPD2, Anti-HB1, Anti-HB2, Anti-HBBD, Anti-HH1, Anti-ICCD, Anti-IVF, Anti-LQT3, Anti-Nav1.5, Anti-PFHB1, Anti-SSS1, Anti-VF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

222kDa

species reactivity

guinea pig, human, mouse, horse, rat, dog, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCN5A(6331)

Immunogen

Synthetic peptide directed towards the C terminal region of human SCN5A

Biochem/physiol Actions

SCN5A is an integral membrane protein and tetrodotoxin-resistant voltage-gated sodium channel subunit. The protein is found primarily in cardiac muscle and is responsible for the initial upstroke of the action potential in an electrocardiogram.The protein encoded by this gene is an integral membrane protein and tetrodotoxin-resistant voltage-gated sodium channel subunit. The encoded protein is found primarily in cardiac muscle and is responsible for the initial upstroke of the action potential in an electrocardiogram. Defects in this gene are a cause of long QT syndrome type 3 (LQT3), an autosomal dominant cardiac disease. Alternative splicing results in two transcript variants encoding separate isoforms which differ by a single amino acid. Mutation nomenclature has been assigned with respect to the longer isoform.

Sequence

Synthetic peptide located within the following region: FTKRVLGESGEMDALKIQMEEKFMAANPSKISYEPITTTLRRKHEEVSAM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianding Cheng et al.
Journal of the American Heart Association, 6(4) (2017-04-05)
We have identified the cardiomyopathy-susceptibility gene vinculin (VCL) mutation M94I may account for a sudden unexplained nocturnal death syndrome (SUNDS) case. We addressed whether VCL common variant D841H is associated with SUNDS. In 8 of 120 SUNDS cases, we detected
Bruno M Fontinha et al.
PLoS biology, 19(1), e3001012-e3001012 (2021-01-08)
Vertebrate behavior is strongly influenced by light. Light receptors, encoded by functional opsin proteins, are present inside the vertebrate brain and peripheral tissues. This expression feature is present from fishes to human and appears to be particularly prominent in diurnal
Jianding Cheng et al.
Scientific reports, 7, 42953-42953 (2017-02-22)
Sudden unexplained nocturnal death syndrome (SUNDS) remains an autopsy negative disorder with unclear etiology. Vinculin (VCL) was linked to sudden arrhythmia death in VCL knockout mice prior to the appearance of cardiomyopathy. We hypothesized VCL mutations underlie risk for SUNDS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service