Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406020

Sigma-Aldrich

Anti-IL2RB antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD122, P70-75

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
FACS
WB
species reactivity:
human
technique(s):
flow cytometry: suitable
western blot: 1 μg/mL
citations:
2

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~61.1 kDa

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IL2RB(3560)

General description

The interleukin 2 receptor, which is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2. The protein encoded by this gene represents the beta subunit and is a type I membrane protein. (provided by RefSeq)

Immunogen

IL2RB (NP_000869.1, 1 a.a. ~ 551 a.a) full-length human protein.

Sequence
MAAPALSWRLPLLILLLPLATSWASAAVNGTSQFTCFYNSRANISCVWSQDGALQDTSCQVHAWPDRRRWNQTCELLPVSQASWACNLILGAPDSQKLTTVDIVTLRVLCREGVRWRVMAIQDFKPFENLRLMAPISLQVVHVETHRCNISWEISQASHYFERHLEFEARTLSPGHTWEEAPLLTLKQKQEWICLETLTPDTQYEFQVRVKPLQGEFTTWSPWSQPLAFRTKPAALGKDTIPWLGHLLVGLSGAFGFIILVYLLINCRNTGPWLKKVLKCNTPDPSKFFSQLSSEHGGDVQKWLSSPFPSSSFSPGGLAPEISPLEVLERDKVTQLLLQQDKVPEPASLSSNHSLTSCFTNQGYFFFHLPDALEIEACQVYFTYDPYSEEDPDEGVAGAPTGSSPQPLQPLSGEDDAYCTFPSRDDLLLFSPSLLGGPSPPSTAPGGSGAGEERMPPSLQERVPRDWDPQPLGPPTPGVPDLVDFQPPPELVLREAGEEVPDAGPREGVSFPWSRPPGQGEFRALNARLPLNTDAYLSLQELQGQDPTHLV

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Rak Cho et al.
Oncology reports, 32(4), 1531-1536 (2014-08-12)
Broussonetia kazinoki (BK) has been used as a traditional medicine to improve vision, as well as for inflammatory and infectious diseases. In the present study, we investigated the effects and molecular mechanism of the ethanolic extract of BK on cell
Diana A Alvarez Arias et al.
Cancer immunology research, 2(3), 207-216 (2014-04-30)
Tumor growth is associated with the inhibition of host antitumor immune responses that can impose serious obstacles to cancer immunotherapy. To define the potential contribution of Qa-1-restricted CD8 regulatory T cells (Treg) to the development of tumor immunity, we studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service