Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402692

Sigma-Aldrich

Monoclonal Anti-APOC3, (C-terminal) antibody produced in mouse

clone 8H7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

APOCIII, MGC150353

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8H7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~34.8 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOC3(345)

General description

Apolipoprotein C3 (APOC3) is a 79 amino acid protein with a molecular weight of 8.8kDa. It is present on high density, low density and triglyceride-rich lipoproteins. The gene encoding this protein is localized on human chromosome 11q23.3.

Immunogen

APOC3 (NP_000031.1, 21 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SEAEDASLLSFMQGYMKHATKTAKDALSSVQESQVAQQARGWVTDGFSSLKDYWSTVKDKFSEFWDLDPEVRPTSAVAA

Biochem/physiol Actions

Apolipoprotein C3 (APOC3) is involved in the metabolism of triglyceride-rich lipoproteins. It may have a role in atherosclerosis. The protein stimulates hypertriglyceridemia (HTG) by inhibiting the activity of lipoprotein lipase and interfering with the binding of APOB or APOE to hepatic receptors. It also promotes very low density lipoprotein (VLDL) assembly and secretion in the liver.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The emerging role of apolipoprotein C-III: beyond effects on triglyceride metabolism.
Luo M and Peng D
Liposome Technology (2016)
Variants in the APOC3 promoter insulin responsive element modulate insulin secretion and lipids in middle-aged men
D. M> Waterworth
Biochim. Biophys. Acta Gen. Subj. (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service