Skip to Content
Merck
All Photos(3)

Key Documents

PLA0273

Sigma-Aldrich

Rabbit anti-c-Jun Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

AP-1, AP1, Jun activation domain binding protein, activator protein 1, c-Jun, enhancer-binding protein AP1, jun oncogene, jun proto-oncogene, p39, proto-oncogene c-Jun, v-jun avian sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
species reactivity:
mouse, human
application:
IHC
IP
WB
technique(s):
immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:2,000-1:10,000
citations:
4

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_002219.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... c-Jun(3725)

Immunogen

The epitope recognized by PLA0273 maps to a region between residue 1 and 40 of human Jun Oncogene using the numbering given in entry NP_002219.1 (GeneID 3725).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

c-Jun is a transcription factor that heterodimerizes with FOS to form the activator protein 1 (AP-1) transcription factor. The JUN gene is the putative transforming gene of avian sarcoma virus 17.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service