Skip to Content
Merck
All Photos(2)

Key Documents

P7521

Sigma-Aldrich

Pro-Arg 4-methoxy-β-naphthylamide acetate salt

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N6O3
CAS Number:
Molecular Weight:
426.51
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

form

powder

color

light orange

storage temp.

−20°C

SMILES string

COc1cc(NC(=O)C(CCCNC(N)=N)NC(=O)C2CCCN2)cc3ccccc13

InChI

1S/C22H30N6O3/c1-31-19-13-15(12-14-6-2-3-7-16(14)19)27-21(30)18(9-5-11-26-22(23)24)28-20(29)17-8-4-10-25-17/h2-3,6-7,12-13,17-18,25H,4-5,8-11H2,1H3,(H,27,30)(H,28,29)(H4,23,24,26)

InChI key

VCBSBIGZWZKYIY-UHFFFAOYSA-N

Substrates

Fluorogenic substrate for cathepsin c

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F A Dolbeare et al.
Clinical chemistry, 23(8), 1485-1491 (1977-08-01)
Enzyme activity can be measured in single cells or in cell suspensions by either static or flow microfluorometry when a fluorogenic substrate is used. We have used amino acid derivatives of arylamines as fluorogenic substrates for tagging cellular proteinases. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service