Skip to Content
Merck
All Photos(1)

Key Documents

P203

Sigma-Aldrich

Phenamil methanesulfonate salt

solid

Synonym(s):

3,5-Diamino-6-chloro-N-[imino(phenylamino)methyl]pyrazinecarboxamide methanesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H12ClN7O · CH3SO3H
CAS Number:
Molecular Weight:
401.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

yellow

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2.5 mg/mL
DMSO: ≥5 mg/mL (with warming)
0.1 M HCl: insoluble
H2O: insoluble

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.Nc1nc(N)c(nc1Cl)C(=O)NC(=N)Nc2ccccc2

InChI

1S/C12H12ClN7O.CH4O3S/c13-8-10(15)19-9(14)7(18-8)11(21)20-12(16)17-6-4-2-1-3-5-6;1-5(2,3)4/h1-5H,(H4,14,15,19)(H3,16,17,20,21);1H3,(H,2,3,4)

InChI key

MHPIZTURFVSLTJ-UHFFFAOYSA-N

General description

Phenamil, a derivative of amiloride is an inhibitor of sodium channels including acid sensing ion channel (ASIC) and epithelial sodium channels (ENaC).

Application

Phenamil methanesulfonate salt has been used:
  • to stimulate adipocyte differentiation in 3T3 cells
  • to test its effect on lateral flagella expression in Vibrio shilonii
  • in nanoparticle preparation to test its effect on osteo/odontoblastic differentiation

Biochem/physiol Actions

Phenamil is an inducer of peroxisome proliferator-activated receptor γ (PPARγ) and inhibits diamine oxidase. By favoring tribbles homolog 3 (Trb3) expression, phenamil holds therapeutic potential to treat pulmonary artery hypertension (PAH) and idiopathic PAH. It is also a bone morphogenetic protein (BMP) activator, which favors osteogenesis and odontogenesis of stem cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Barbry et al.
Biochemistry, 28(9), 3744-3749 (1989-05-02)
The existence of distinct forms of the epithelium Na+ channel that differ in their sensitivity to amiloride has been repeatedly suggested by physiological data. The biochemical basis for these differences was analyzed by using phenamil, the most potent inhibitor known
Yael González et al.
Canadian journal of microbiology, 61(3), 183-191 (2015-02-03)
In this study, we show the induction of lateral flagella by the action of the sodium channel blocker phenamil, in the marine bacterium Vibrio shilonii, a coral pathogen that causes bleaching. We analyzed the growth and morphology of cells treated
J L Garvin et al.
The Journal of membrane biology, 87(1), 45-54 (1985-01-01)
Several new amiloride analogues and two reported photoaffinity analogues were tested for irreversible inhibition of short-circuit current, Isc, in toad bladder. Bromoamiloride, a photoaffinity analogue, induced 40% irreversible inhibition at 500 microM after irradiation with ultraviolet light greater than or
N Voilley et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(1), 247-251 (1994-01-04)
Water balance in the lung is controlled via active Na+ and Cl- transport. Electrophysiological measurements on lung epithelial cells demonstrated the presence of a Na+ channel that is inhibited by amiloride (K0.5 = 90 nM) and some of its derivatives
Mun Chun Chan et al.
Molecular and cellular biology, 31(3), 517-530 (2010-12-08)
Pulmonary artery hypertension (PAH) is characterized by elevated pulmonary artery resistance and increased medial thickness due to deregulation of vascular remodeling. Inactivating mutations of the BMPRII gene, which encodes a receptor for bone morphogenetic proteins (BMPs), are identified in ∼60%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service