Skip to Content
Merck
All Photos(2)

Key Documents

L2643

Sigma-Aldrich

α-Lactose monohydrate

suitable for cell culture, BioReagent

Synonym(s):

β-D-Gal-(1→4)-α-D-Glc, 4-O-β-D-Galactopyranosyl-α-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

biological source

bovine milk

product line

BioReagent

Assay

≥99% total lactose basis (GC)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.05% Glucose
≤4% β-lactose

solubility

water: 100 mg/mL, clear, colorless

SMILES string

[H]O[H].OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11.H2O/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17;/h3-20H,1-2H2;1H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+;/m1./s1

InChI key

WSVLPVUVIUVCRA-KPKNDVKVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Lactose monohydrate has been used:
  • as a component of fraction B of diluent M1 for dilution of ram semen
  • as a component of Platz diluent variant (PDV) for dilution of bottlenose dolphin semen
  • in the preparation of HyQ PF-Chinese hamster ovary (CHO) disaccharide media for the adaptation of CHO cell lines
  • for the preparation of protein-free chemically defined cell culture medium (PFCDM) for the adaptation of human embryonic kidney 293 (HEK293) cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Robeck et al.
Biology of reproduction, 70(5), 1340-1348 (2004-01-09)
Research was conducted to develop an effective method for cryopreserving bottlenose dolphin (Tursiops truncatus) semen processed immediately after collection or after 24-h liquid storage. In each of two experiments, four ejaculates were collected from three males. In experiment 1, three
A Thiel et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 125, 528-539 (2019-02-06)
3-NOP (3-nitrooxypropanol) reduces enteric methane formation in ruminants. A series of ADME studies in rats, lactating goats and beef cattle was performed. 3-NOP was entirely absorbed from the GIT of rats: approximately 75% of the administered 3-NOP was eliminated as
L Anel et al.
Theriogenology, 60(7), 1293-1308 (2003-09-27)
Glycerol has been the most widely used cryopreservation agent for spermatozoa and a wide range of factors affect its action on sperm viability and fertilizing capacity. We tested three methods for freezing ram semen packed in 0.25 ml straws (final
Ling Zhang et al.
Theranostics, 10(24), 11339-11358 (2020-10-13)
Background: TSTA3 gene encodes an enzyme responsible for synthesis of GDP-L-fucose as the only donor in fucosylation. This study was designed to explore clinical value, function and underlying mechanism of TSTA3 in the development of esophageal squamous cell carcinoma (ESCC).
Dawn Sow Zong Leong et al.
Scientific reports, 7, 45216-45216 (2017-03-31)
Mammalian cells are generally considered to be unable to utilize polysaccharides for cell growth because the phospholipid bilayer in the cell membrane has very low permeability to sugars. With the recent discovery of the only known animal disaccharide transporter, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service