Skip to Content
Merck
All Photos(5)

Key Documents

HPA026808

Sigma-Aldrich

Anti-PRRX2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C17orf33, Anti-G-CSF, Anti-GCSF, Anti-MGC45931, Anti-PMX2, Anti-PRX2, Anti-paired related homeobox 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRRX2(51450)

General description

Paired related homeobox 2 (PRRX2) is a member of PRRX gene family, which is encoded by a gene mapped to human chromosome 9q34.1. It is primarily expressed in proliferating fetal fibroblasts and developing dermis. Apart from this, reverse transcription polymerase chain reaction (RT-PCR) analysis of human tissue revealed the presence of PRRX2 in the kidney and lung.

Immunogen

paired related homeobox 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Paired related homeobox 2 (PRRX2), along with homeobox B13(HOXB13), plays a vital role in scarless fetal skin regeneration. Mutations in the PRRX2 gene might lead to (NAFD) syndrome. Experimental studies on NIH-3T3cells (mouse embryonic fibroblast cell line) shows that conserved PRX domain of PRRX2 facilitates cell-specific and promoter-dependent transcriptional regulation. In rat, PRRX2 facilitates embryonic pituitary development by promoting vasculogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70972

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human PRRX1 and PRRX2 genes: cloning, expression, genomic localization, and exclusion as disease genes for Nager syndrome.
Norris RA
Mammalian Genome, 11(11), 1000-1005 (2000)
Modulation of the human homeobox genes PRX-2 and HOXB13 in scarless fetal wounds.
Stelnicki EJ
The Journal of Investigative Dermatology, 111(1), 57-63 (1998)
Yu-Lin Juang et al.
Molecular carcinogenesis, 55(12), 2247-2259 (2016-01-30)
TGF-β and cancer progression share a multifaceted relationship. Despite the knowledge of TGF-β biology in the development of cancer, several factors that mediate the cancer-promoting role of TGF-β continue to be identified. This study aimed to identify and characterise novel
Identification of domains mediating transcription activation, repression, and inhibition in the paired-related homeobox protein, Prx2 (S8).
Norris RA, Kern MJ
Dna and Cell Biology, 20(2), 89-99 (2001)
SPOCK1 Is a Novel Transforming Growth Factor-?-Induced Myoepithelial Marker That Enhances Invasion and Correlates with Poor Prognosis in Breast Cancer.
Fan LC
PLoS ONE, 11(9) (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service