Skip to Content
Merck
All Photos(2)

Key Documents

HPA021221

Sigma-Aldrich

Anti-MMP8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-MMP-8, Anti-Matrix metalloproteinase-8, Anti-Neutrophil collagenase, Anti-PMNL collagenase, Anti-PMNL-CL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

KWERTNLTYRIRNYTPQLSEAEVERAIKDAFELWSVASPLIFTRISQGEADINIAFYQRDHGDNSPFDGPNG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP8(4317)

General description

The gene MMP8 (matrix metalloproteinase-8) is mapped to human chromosome 11q22.2-q22.3. It belongs to the MMP family of proteins.

Immunogen

Neutrophil collagenase Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MMPs (matrix metalloproteinases) are zinc-dependent endopeptidases which are mainly responsible for degradation of extracellular matrix components. MMP8 is a neutrophilic collagenase. It degrades fibrillar collagen type I, type II and III, serpins and α-2 macroglobulin. It can also degrade apolipoprotein A-1, leading to high accumulation of cholesterol in the vessel walls.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75159

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Şurlin et al.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie, 55(3 Suppl), 1137-1141 (2015-01-22)
There is scientific data to support the existence of a two-way relationship between diabetes and periodontitis, with diabetes increasing the risk for periodontitis, and periodontal inflammation negatively affecting the diabetic status. Our study aims to investigate the expression of MMP-7
Sébastien Lenglet et al.
Mediators of inflammation, 2013, 659282-659282 (2013-02-01)
Plaque rupture is the main cause of acute myocardial infarction and stroke. Atherosclerotic plaques have been described to be vulnerable and more prone to rupture when they are characterized by thin, highly inflamed, and collagen-poor fibrous caps and contain elevated
Aino Salminen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1435-1445 (2015-01-01)
Various cell types in atherosclerotic lesions express matrix metalloproteinase (MMP)-8. We investigated whether MMP-8 affects the structure and antiatherogenic function of apolipoprotein (apo) A-I, the main protein component of HDL particles. Furthermore, we studied serum lipid profiles and cholesterol efflux
Timo Rath et al.
PloS one, 9(12), e115847-e115847 (2014-12-30)
Cystic Fibrosis (CF) lung disease is characterized by progressively declining lung function and represents a major factor contributing to the high morbidity and mortality associated with CF. However, apart from spirometry, respiratory disease surrogate markers reliably indicating CF lung disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service