GE18117263
Fingertight Connector 1/16" Male
ÄKTA®express, Cytiva 18117263
All Photos(1)
About This Item
UNSPSC Code:
41115705
NACRES:
NB.43
Recommended Products
material
black PEEK
feature
Connections: Fingertight 1/16"
Wetted Material: Polyetheretherketone (PEEK)
ÄKTA®express
manufacturer/tradename
Cytiva 18117263
compatibility
For use with: Tubing O.D. 1/16"
Legal Information
ÄKTA is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)