Key Documents
EP0030076249
Eppendorf® Empty Box 2.0 for epT.I.P.S.® Reloads
new
for tips 10 – 300 µL
About This Item
UNSPSC Code:
41121600
volume:
10—300 μL
feature:
for tips 10 – 300 µL
packaging:
pkg of 1 ea
manufacturer/tradename:
Eppendorf® 0030076249
feature
for tips 10 – 300 µL
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 0030076249
volume
10—300 μL
General description
epT.I.P.S.® Box 2.0 for epT.I.P.S.® Reloads, empty box, does not include tips, Eppendorf Quality, 10 – 300 µL
Application
General liquid handling
Features and Benefits
  • epT.I.P.S.® Box 2.0 – empty reusable boxes for tip volumes of 10 µL - 5 mL
  • epT.I.P.S.® Box 2.0 – all three sizes are availabe in Eppendorf Quality
  • Autoclavable at 121 °C (20 min.) for subsequent use
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)