EP0030000870
Eppendorf® epT.I.P.S.® standard
volume range 2-200 μL, yellow polypropylene, pack of 1000 ea (2 bags × 500 tips)
Synonym(s):
pipetting tips
About This Item
Recommended Products
material
yellow polypropylene
sterility
non-sterile
feature
Eppendorf Quality™
packaging
pack of 1000 ea (2 bags × 500 tips)
manufacturer/tradename
Eppendorf® 0030000870
availability
available only in (India)
maximum volume
200 μL
volume range
2-200 μL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Original and high-quality pipettes packed in resealable bags
- Tips are autoclave-friendly at 121°C for 20 mins
- The fine retracted or extended tips enable simpler ways to work with small sample volumes
- Universal tips that can be used with Eppendorf pipettes and pipettes from other manufacturers
- Disposable racks with a slim design, reclosable and easy stackability
- The cone shape of the tips ensures optimal pipette fit with Eppendorf pipettes
- Fine graduations enable visual examination of the pipetting volume even easier
- Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service