Skip to Content
Merck
All Photos(1)

Documents

EHU076051

Sigma-Aldrich

MISSION® esiRNA

targeting human NOX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCACAACCTCACCTTCCACAAGCTGGTGGCCTATATGATCTGCCTACATACAGCTATTCACATCATTGCACACCTGTTTAACTTTGACTGCTATAGCAGAAGCCGACAGGCCACAGATGGCTCCCTTGCCTCCATTCTCTCCAGCCTATCTCATGATGAGAAAAAGGGGGGTTCTTGGCTAAATCCCATCCAGTCCCGAAACACGACAGTGGAGTATGTGACATTCACCAGCATTGCTGGTCTCACTGGAGTGATCATGACAATAGCCTTGATTCTCATGGTAACTTCAGCTACTGAGTTCATCCGGAGGAGTTATTTTGAAGTCTTCTGGTATACTCACCACCTTTTTATCTTCTATATCCTTGGCTTAGGGATTCACGGCATTGGTGGAATTGTCCGGGGTCAAACAGAGGAGAGCATGAATGAGAGTCATCCTCGCAAGTGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerco den Hartog et al.
PLoS pathogens, 12(1), e1005382-e1005382 (2016-01-14)
Generation of reactive oxygen species (ROS) during infection is an immediate host defense leading to microbial killing. APE1 is a multifunctional protein induced by ROS and after induction, protects against ROS-mediated DNA damage. Rac1 and NAPDH oxidase (Nox1) are important
The nuclear receptor NOR-1 modulates redox homeostasis in human vascular smooth muscle cells.
Judith Alonso et al.
Journal of molecular and cellular cardiology, 122, 23-33 (2018-08-11)
H-P Wang et al.
European review for medical and pharmacological sciences, 20(21), 4474-4481 (2016-11-23)
Reactive oxygen species (ROS) generated by endogenous metabolic enzymes are involved in a variety of pathology processes, including cancer. In particular, superoxide-generating NADPH oxidase 1 (Nox1), a member of Nox enzyme family, is highly expressed in the colon tissue and
Jin-Woo Jeong et al.
International journal of molecular sciences, 20(6) (2019-03-25)
Excessive bone resorption by osteoclasts causes bone loss-related diseases and reactive oxygen species (ROS) act as second messengers in intercellular signaling pathways during osteoclast differentiation. In this study, we explored the protective effects of fermented oyster extract (FO) against receptor
So Yeong Park et al.
International journal of molecular sciences, 21(20) (2020-10-22)
Y-27632 is known as a selective Rho-associated coiled coil-forming kinase (ROCK) inhibitor. Y-27632 has been shown to induce neurite outgrowth in several neuronal cells. However, the precise molecular mechanisms linking neurite outgrowth to Y-27632 are not completely understood. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service