CLS3822
Corning® 384 well microplate, low volume
Tissue Culture (TC)-treated surface, black polystyrene, round bottom, sterile, lid, pack of 10
Synonym(s):
384 multiwell plates
About This Item
black polystyrene
round bottom
skirt
Recommended Products
material
black bottom
black polystyrene
round bottom
sterility
sterile
feature
lid
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3822
maximum volume
50 μL
size
384 wells
working volume
40 μL
color
black
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service