Skip to Content
Merck
All Photos(1)

Key Documents

CLS3690

Corning® 96 Well Half-Area Microplate

flat bottom clear, polystyrene, High, bag of 25 inner package, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear
polystyrene
round wells
size:
96 wells
Pricing and availability is not currently available.

material

flat bottom clear
polystyrene
round wells

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25 inner package

manufacturer/tradename

Corning 3690

maximum volume

190 μL

size

96 wells

well D

10.7 mm

working volume

125 μL

color

clear

suitability

suitable for (cell cuture)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate without lids conforms to standard microplate footprint and dimensions.
  • High binding surface is capable of binding medium (>10kD) and large biomolecules with ionic groups and hydrophobic regions.
  • Binding capacity is approximately 400 to 500ng IgG/cm².
  • Ideal for immunoassays.
  • Recommended working volume of up to 25 to 125 μL.
  • Conforms to standard microplate footprint and dimensions.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service