Skip to Content
Merck
All Photos(1)

Key Documents

CLS3524

Corning® Costar® TC-Treated Multiple Well Plates

size 24 wells, polystyrene plate, flat bottom wells, case of 100 (individually wrapped), sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NB.13
material:
clear bottom
flat bottom wells
polystyrene plate
round clear wells
size:
24 wells
well volume:
3.4 mL
well working volume:
0.38-0.57 mL
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
skirt
Pricing and availability is not currently available.

material

clear bottom
flat bottom wells
polystyrene plate
round clear wells

sterility

sterile

feature

lid
skirt

packaging

case of 100 (individually wrapped)

manufacturer/tradename

Corning 3524

size

24 wells

well volume

3.4 mL

well working volume

0.38-0.57 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Features and Benefits

  • Specially treated to promote optimalcell attachment.
  • Ensured sterility through gammairradiation.
  • Nonreversible lids with condensationrings to reduce contamination
  • Certified non-pyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service