MFE-296
98031101, human endometrium, Epithelial
Synonym(s):
MFE296 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
MFE-296, 98031101
biological source
human endometrium
growth mode
Adherent
karyotype
Near Tetraploid
morphology
Epithelial
products
Not specified
receptors
Androgen receptor
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human caucasian endometrial adenocarcinoma
Cell Line Description
MFE-296 was obtained from a moderately differentiated human endometrial adenocarcinoma. The cells were shown to express functionally intact androgen receptors. Proliferation was inhibted by progestin R5020 and dihydrotestosterone (DHT), the latter one being antagonised by the antiandrogen Casodex. MFE-296 express vimentin and the cytokeratins 7, 8, 18 and 19. Karyotype analysis revealed near tetraploidy for most cells. Further more, the cells have been shown to be tumorigenic in nude mice.
Application
Tumour biology studies, receptor analysis
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 8,13
D16S539: 11,14
D5S818: 11,13
D7S820: 8,10
THO1: 6,9.3
TPOX: 8
vWA: 14,18
CSF1PO: 10
D13S317: 8,13
D16S539: 11,14
D5S818: 11,13
D7S820: 8,10
THO1: 6,9.3
TPOX: 8
vWA: 14,18
Culture Medium
MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 i.e. seeding 1-2 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service