Skip to Content
Merck
All Photos(1)

Key Documents

CHO/dhFr-

94060607, hamster ovary, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CHO/dhFr-, 94060607

biological source

hamster ovary

growth mode

Adherent

karyotype

Modal no. 20

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese ovary

Cell Line Description

The CHO/dhFr- cell line lacks the enzyme dihydrofolate reductase (DHFR) which is necessary for purine synthesis. In the absence of exogenous purines, this enzyme is required for growth. Hence the CHO/dhFr- cell line has growth requirements for hypoxanthine (or adenine), glycine and thymidine. However, the cells will continue to grow without the addition of hypoxanthine and thymidine because the cells can obtain sufficient from the medium. α MEM minus does not contain the ribonucleosides or the deoxyribonucleosides, and will not support the growth of CHO/dhFr- or its serum free adaptation as neither contains the gene to produce DHFR. (See attached data sheet).

Application

CHO/dhFr- has been used to study the relation between local chromatin mobility and transcriptional activity. It has also been used as a transfection lineage and also as a negative control in cytofluorometry experiments.

Culture Medium

IDMEM + 2mM Glutamine + 0.1mM Hypoxanthine + 0.01mM Thymidine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service