Skip to Content
Merck
All Photos(1)

Documents

ND7/23 Cell Line

92090903, Mouse x Rat Hybridoma nerve, Neuronal

Synonym(s):

ND7/23 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ND7/23 Cell Line, 92090903, Mouse neuroblastoma X Rat neuron hybrid

biological source

Mouse x Rat Hybridoma nerve

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat neurone hybrid

Cell Line Description

Mouse neuroblastoma (N18 tg 2) x rat dorsal root ganglion neurone hybrid cell line produced by PEG mediated cell fusion.

Application

Mouse neuroblastoma (N18 tg 2) and rat dorsal root glanglia produced by PEG-mediated fusion.
Recommended culture medium: DMEM + 2 mM L-Glutamine + 10% FBS
Subculture: Split at 70-80% confluency, approx. 1:3 to 1:6, seed at 1-3x104 viable cells/cm2 at 37°C and 5% CO2. Dissociate cells by aspiration or gentle tapping. Cells are lightly adherent, no trypsinization is necessary.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 by either aspirating or gently tapping the cells loose; 5% CO2; 37°C. Cells are lightly attached therefore use of trypsin is unnecessary.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service