Skip to Content
Merck
All Photos(1)

Key Documents

OKT3

Reactivity: Anti-CD3, human, 86022706, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 86022706-DNA-5UG
pkg of vial of cells 86022706-1VL

ImmunoDon

CAF1 mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-CD3, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8 x Mouse CAF1 spleen

Cell Line Description

The antibody is directed against the CD3 antigen on human T cells.

Immunogen

Human peripheral blood lymphocytes

Culture Medium

IDMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells may be slow to grow upon resuscitation and may take up to 7 days before they are ready for subculture.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service