Skip to Content
Merck
All Photos(1)

Documents

C4866

Sigma-Aldrich

Cardiogenol C hydrochloride

≥97% (HPLC), solid

Synonym(s):

4-(2-Hydroxyethylamino)-2-(4-methoxyphenylamino)-pyrimidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O2 ·HCl
CAS Number:
Molecular Weight:
296.75
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

solid

mol wt

296.75

packaging

pkg of 5 mg

solubility

H2O: soluble 10 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].COc1ccc(Nc2nccc(NCCO)n2)cc1

InChI

1S/C13H16N4O2.ClH/c1-19-11-4-2-10(3-5-11)16-13-15-7-6-12(17-13)14-8-9-18;/h2-7,18H,8-9H2,1H3,(H2,14,15,16,17);1H

InChI key

QQAHYSZVJLHCNA-UHFFFAOYSA-N

Biochem/physiol Actions

Cardiogenol C hydrochloride is a cardiomyogenesis inducer in embryonic stem cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Wu et al.
Journal of the American Chemical Society, 126(6), 1590-1591 (2004-02-12)
A phenotypic cell-based screen of a large combinatorial chemical library led to the identification of a class of diaminopyrimidine compounds (cardiogenol A-D) which can selectively and efficiently induce mouse embryonic stem cells (ESCs) to differentiate into cardiomyocytes. ESC-derived cardiomyocytes were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service