C4567
Utility carriers, Nalgene®
L × W × H 15 in. (381 mm) × 14 in. (356 mm) × 10 1/2 in. (267 mm)
Synonym(s):
laboratory storage, plastic storage box
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
high-density polyethylene
feature
Holds up to 6 x 1000 mL bottles
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 7131-0001
L × W × H
15 in. (381 mm) × 14 in. (356 mm) × 10 1/2 in. (267 mm)
Looking for similar products? Visit Product Comparison Guide
General description
Resilient. Molded-in center divider and handle. Stackable when empty for compact storage.
With removable foam inserts to protect six bottles. With inserts removed, carrier accommodates Unwire test tube racks, square bottles etc. Sturdy enough to hold six full 1-liter bottles.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service