Skip to Content
Merck
All Photos(3)

Key Documents

AV51196

Sigma-Aldrich

Anti-MAS1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-MAS, Anti-MAS1 oncogene, Anti-MGC119966

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAS1(4142)

Related Categories

Immunogen

Synthetic peptide directed towards the middle region of human MAS1

Application

Anti-MAS1 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

MAS1 is a transmembrane G-protein-coupled receptor for angiotensin-1 to -7. It activates the phospholipase C pathway and has a role in smoothe muscle relaxation, hypotension and cardioprotection.

Sequence

Synthetic peptide located within the following region: RPKYQSALVCALLWALSCLVTTMEYVMCIDREEESHSRNDCRAVIIFIAI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcio Augusto Fressatto de Godoy et al.
International journal of hypertension, 2012, 121740-121740 (2011-12-14)
Ang-(1-7) is produced via degradation of Ang II by the human angiotensin converting enzyme, also known as ACE2. In the cardiovascular system, Ang-(1-7) has been shown to produce effects that are opposite to those of Ang II. These include smooth
Enéas R M Gomes et al.
International journal of hypertension, 2012, 493129-493129 (2012-04-21)
The Renin-Angiotensin System (RAS) acts at multiple targets and has its synthesis machinery present in different tissues, including the heart. Actually, it is well known that besides Ang II, the RAS has other active peptides. Of particular interest is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service