Skip to Content
Merck
All Photos(2)

Key Documents

537705

Sigma-Aldrich

5-Methyl-2-hexanone

99%

Synonym(s):

MIAK, Isobutylacetone, Isopentyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2COCH3
CAS Number:
Molecular Weight:
114.19
Beilstein:
506163
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
145 °C (lit.)
vapor pressure:
4.5 mmHg ( 20 °C)

vapor density

3.94 (vs air)

Quality Level

vapor pressure

4.5 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

797 °F

expl. lim.

1.35-8.2 %, 93 °F

impurities

≤0.05% (water)

refractive index

n20/D 1.406 (lit.)

bp

145 °C (lit.)

mp

-74 °C

solubility

water: soluble 5.4 g/L at 25 °C

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCC(C)=O

InChI

1S/C7H14O/c1-6(2)4-5-7(3)8/h6H,4-5H2,1-3H3

InChI key

FFWSICBKRCICMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methyl-2-hexanone (methyl isoamyl ketone, MIAK) is an aliphatic ketone. Its ability to elicit germination response in uredospores of certain species of rust has been investigated. The near-UV absorption cross section and absorption spectrum of MIAK were measured and employed to determine its photolysis lifetime. The dynamics of the Norrish Type-II reaction of MIAK has been studied using femtosecond time-resolved mass spectrometry.

Application

5-Methyl-2-hexanone may be used to synthesize N,N′-bis(1,4-dimethylpentyl)-p-phenylenediamine (BMPPD) via reductive alkylation of p-nitroaniline in the presence of copper-based catalysts.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Germination responses of several species of rust spores to 5-methyl-2-hexanone, isomers of ionone, and other structurally related flavor compounds.
French RC.
Journal of Agricultural and Food Chemistry, 31(2), 423-427 (1983)
The near-UV absorption cross sections for several ketones.
Yujing M and Mellouki A.
Journal of Photochemistry and Photobiology A: Chemistry, 134(1), 31-36 (2000)
Study on Copper-based Catalysts for Synthesis of N,N'-bis(1,4-dimethylpentyl)-p-phenylenediamine from Reductive Alkylation of p-Nitroaniline with 5-Methyl-2-hexanone
Pan ZD, et al.
Catalysis Letters, 122(1-2), 115-120 (2008)
Femtosecond Dynamics of Norrish Type-II Reactions: Nonconcerted Hydrogen-Transfer and Diradical Intermediacy.
Feyter SD, et al.
Angewandte Chemie (International Edition in English), 39(1), 260-263 (2000)
G V Katz et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 6(3), 498-505 (1986-04-01)
Rats were exposed by inhalation, 6 hr/day, 5 days/week, to target vapor concentrations of 2000, 1000, or 0 ppm of methyl isoamyl ketone (MIAK) for 12 exposures spanning 16 days, and 2000, 1000, 200, or 0 ppm for 69 exposures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service