Skip to Content
Merck
All Photos(1)

Key Documents

Y0000768

Etodolac for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Etodolac, 1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, Etodolic acid, NSC 282126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3
CAS Number:
Molecular Weight:
287.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etodolac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCc1cccc2c3CCOC(CC)(CC(O)=O)c3[nH]c12

InChI

1S/C17H21NO3/c1-3-11-6-5-7-12-13-8-9-21-17(4-2,10-14(19)20)16(13)18-15(11)12/h5-7,18H,3-4,8-10H2,1-2H3,(H,19,20)

InChI key

NNYBQONXHNTVIJ-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etodolac for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX-1 and COX-2.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yael Goldfarb et al.
Annals of surgery, 253(4), 798-810 (2011-04-09)
Surgical procedures, including primary tumor resection, have been suggested to suppress immune competence and to promote postoperative infections and cancer metastasis. Catecholamines and prostaglandins were recently implicated in these processes, and in directly promoting tumor angiogenesis and invasion. To examine
N Zvaifler
Clinical rheumatology, 8 Suppl 1, 43-53 (1989-03-01)
Etodolac (Lodine, Ramodar, Ultradol), an anti-inflammatory, analgesic agent, is the first of a new class of nonsteroidal anti-inflammatory drugs (NSAIDs), the pyranocarboxylic acids. A review of the literature on numerous clinical studies showed that etodolac (200 to 600 mg/day) is
S Lynch et al.
Drugs, 31(4), 288-300 (1986-04-01)
Etodolac is a new non-steroidal agent (NSAID) with anti-inflammatory and analgesic activity. It has been studied in rheumatoid arthritis, osteoarthritis, and pain resulting from minor surgery. In the latter setting, etodolac is at least as effective as aspirin in terms
D R Brocks et al.
Clinical pharmacokinetics, 26(4), 259-274 (1994-04-01)
Etodolac is a chiral nonsteroidal anti-inflammatory drug (NSAID) that is marked as the racemate. Currently, the drug is available in several countries for the treatment of arthritis and the alleviation of pain. Etodolac possesses several unique disposition features mainly due
Etodolac: the chemistry, pharmacology, metabolic disposition, and clinical profile of a novel anti-inflammatory pyranocarboxylic acid.
L G Humber
Medicinal research reviews, 7(1), 1-28 (1987-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service