Skip to Content
Merck

SYNPLE-B032

Reagent-cartridge Nosyl Protection

Synple

Synonym(s):

Synple 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


reaction suitability

reaction type: Amine Protection

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

General description

Reagent-cartridge Nosyl Protection is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the Nosyl protection of primary and secondary free amines. The cartridge contains all necessary reagents to perform the protection of alkyl amines.

Scale: up to 0.8 mmol

Compatible substrates: primary alkyl amines, secondary alkyl amines

Cartridge contents:
  • Silica supported triethylamine and o-Ns-Cl (for protection)
  • Silica supported Trisamine (for purification)
Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
  • Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
  • Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety – all chemicals are contained in the system, minimizing the user’s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system – simple installation
  • Reaction temperatures from 25 °C – 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG


pictograms

Skull and crossbonesCorrosion

signalword

Danger

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library