Skip to Content
Merck
All Photos(1)

Key Documents

PHR1414

Supelco

Methyl Stearate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Methyl stearate, Methyl octadecanoate, Stearic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786213
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M1770200
traceable to USP 1437508

API family

methyl stearate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methyl stearate is a saturated methyl ester and a representative key molecule of biodiesel fatty acid methyl esters (FAMEs) produced from vegetable oils. It occurs naturally in clove oil.

Application

Methyl Stearate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by gas-chromatography (GC) coupled to flame-ionization detector (FID).[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1964 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl Stearate
USP42-NF37
United States Pharmacopeia/National Formulary, 6180-6180 (2013)
Christophe Capallere et al.
Toxicology in vitro : an international journal published in association with BIBRA, 53, 45-56 (2018-07-28)
In recent years, in vitro skin models combining cell biology and tissue engineering have been developed in order to replace animal models for toxicological studies and to serve as research support to better understand skin biology. This study reports the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service