Skip to Content
Merck
All Photos(1)

Key Documents

O0240000

Oxolinic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO5
CAS Number:
Molecular Weight:
261.23
Beilstein:
620635
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxolinic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1C=C(C(O)=O)C(=O)c2cc3OCOc3cc12

InChI

1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)

InChI key

KYGZCKSPAKDVKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxolinic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drug therapy reviews: oxolinic acid.
R Gleckman et al.
American journal of hospital pharmacy, 36(8), 1077-1079 (1979-08-01)
Guyue Cheng et al.
Analytical and bioanalytical chemistry, 406(30), 7899-7910 (2014-10-31)
Fluoroquinolones (FQNs) are broad-spectrum antibacterial agents widely used in animal husbandry and aquaculture. The residues and antimicrobial resistance of such antibiotics are a major public health concern. To realize multianalyte detection of FQN residues, a genetically modified bacterium, Escherichia coli
Magda Targa Martins et al.
Talanta, 129, 374-383 (2014-08-17)
In routine laboratory work, screening methods for multiclass analysis can process a large number of samples in a short time. The main challenge is to develop a methodology to detect as many different classes of residues as possible, combined with
Maki Kanda et al.
Journal of AOAC International, 98(1), 230-247 (2015-04-11)
Residues of 37 polar veterinary drugs belonging to six families (quinolones, tetracyclines, macrolides, lincosamides, sulfonamides, and others) in livestock and fishery products were determined using a validated LC-MS/MS method. There were two key points in sample preparation. First, extraction was
Diego G Rocha et al.
Journal of chromatography. A, 1379, 83-91 (2015-01-13)
This work involved the optimization and validation of two methods according to the Commission Decision 2002/657/EC directives for determining fluoroquinolones residues in samples of poultry muscle and kidney: ciprofloxacin, danofloxacin, difloxacin, enrofloxacin, flumequine, marbofloxacin, nalidixic acid, norfloxacin, ofloxacin, oxolinic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service