Skip to Content
Merck
All Photos(1)

Key Documents

NIST1885B

Portland cement

NIST® SRM® 1885a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 vials x 5 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1885b comprises five vials, each containing around ~5 g of Portland cement ground to pass through a 75 µm (No. 200) sieve, and each vial is sealed in a foil pouch.For more information, please refer to the COA and SDS.

SRM 1885B_cert

SRM 1885B_SDS

Application

The Standard Reference Material (SRM) is a Type I-IIPortland cement primarily intended to evaluate the methods for analyzing thecements and materials of similar matrix.

Features and Benefits

  • Accompanied by NIST Certified mass fraction values for variousconstituents on an as-received basis, that have metrological traceability tothe SI unit of mass.
  • Information values are listed for Loss on Ignition (LOI) between ambienttemperature and 45 º C, along with the total analyzed constituents.
  • Expiration details, instructions for use, handling, and storage areprovided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminum Oxide (Al2O3) , Ferric Oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulfur trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O) , Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Zinc oxide (ZnO), Manganese(III) oxide (Mn2O3), Chromium(III) oxide (Cr2O3), Strontium oxide (SrO) , Barium oxide (BaO)

Reference/Informational value is provided for the following analytes.
Sulfide sulfur, Insoluble residue, Free CaO, Fluoride (F-), Chlorine (Cl)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service