Skip to Content
Merck
All Photos(1)

Key Documents

68405

Supelco

MCPA-(methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-2-(methyl-d3)phenoxyacetic acid, MCPA-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D3H6ClO3
Molecular Weight:
203.64
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

Related Categories

General description

MCPA-(methyl-d3) is an isotopically labeled compound of the pesticide 4-chloro-2-methylphenoxy acetic acid (MCPA). MCPA is a widely used acidic herbicide which is soluble in water and highly effective even at low concentrations.

Application

MCPA-(methyl-d3) may be used as a internal standard for the determination of the target pesticide in surface water by on-line solid-phase extraction−liquid chromatography-tandem mass spectrometry (SPE−LC−MS/MS). It may also be used as an internal standard for the quantification of the target analyte in soils by liquid chromatography-tandem mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and rapid analytical methodology based on liquid chromatography-tandem mass spectrometry for monitoring pesticide residues in soils from Argentina
De Geronimo, et al.
Analytical Methods : Advancing Methods and Applications, 7(22), 9504-9512 (2015)
Hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry for acidic herbicides and metabolites analysis in fresh water.
Fauvelle V, et al.
Environmental Science and Pollution Research International, 22(6), 3988-3996 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service