52424
Potato Infusion Powder
suitable for microbiology, Component of media such as Potato Dextrose Agar (PDA)
Synonym(s):
PI Powder
About This Item
biological source
potato
Quality Level
sterility
non-sterile
form
powder
shelf life
limited shelf life, expiry date on the label
composition
amino-N, ≥2.0%
nitrogen content, ≥3.0%
storage condition
dry at room temperature
ign. residue
≤22%
loss
≤8.0% loss on drying
pH
5.0-7.0 (25 °C, 2% in H2O)
solubility
H2O: 2%, clear to turbid (light yellow and light brown-yellow and light brown)
application(s)
microbiology
Related Categories
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service