Skip to Content
Merck
All Photos(2)

Key Documents

45314

Supelco

Thiofanox

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O2S
CAS Number:
Molecular Weight:
218.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(ON=C(C(C)(C)C)CSC)=O

InChI

1S/C9H18N2O2S/c1-9(2,3)7(6-14-5)11-8(12)13-10-4/h10H,6H2,1-5H3/b11-7+

InChI key

AGBTZYDUBVIKEZ-YRNVUSSQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of carbamate insecticide thiofanox in rats.
B H Chin et al.
Journal of agricultural and food chemistry, 28(6), 1085-1090 (1980-11-01)
Anticholinesterase effects of carbamate insecticide thiofanox and its metabolites in rats.
B H Chin et al.
Journal of agricultural and food chemistry, 28(6), 1327-1330 (1980-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service