Skip to Content
Merck
All Photos(2)

Documents

44310

Sigma-Aldrich

AmberChrom 1X4 Ion Exchange Resin

chloride form, 50-100 mesh

Synonym(s):

Dowex 1X4, Cl(−)-form, strongly basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

product name

AmberChrom 1X4 chloride form, chloride form, 50-100 mesh

Quality Level

description

cross-linkage 4%

form

beads

parameter

66 °C OH- form max. temp.
99 °C Cl- form max. temp.

moisture

>50%

technique(s)

LPLC: suitable

loss

~55% loss on drying, 110 °C

matrix

styrene-divinylbenzene (gel)

matrix active group

SAX

particle size

50-100 mesh

operating pH

0-14

capacity

1.0 meq/mL capacity

separation technique

anion exchange

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dowex 1X4 chloride form is a strongly basic (type I) anion exchange resin for fine pharmaceuticals. Dowex 1X4 ion exchangers have been used to remove iodinated contrast media (ICM), water pollutants, from the water phase, suggesting a removal method for use in aquatic environments. Dowex 1X4 has also been used to determine partitioning of heavy metals (Ca, Mg, Fe and Zn) in cow milk.
Strongly basic (type I) anion exchange resin for fine pharmaceuticals; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle Caetano et al.
Journal of colloid and interface science, 338(2), 402-409 (2009-08-15)
The removal of phenol from aqueous solution was evaluated by using a nonfunctionalized hyper-cross-linked polymer Macronet MN200 and two ion exchange resins, Dowex XZ (strong anion exchange resin) and AuRIX 100 (weak anion exchange). Equilibrium experimental data were fitted to
Pawel Pohl et al.
Talanta, 71(2), 715-721 (2007-02-15)
An operationally defined fractionation protocol was developed to study the partitioning of Ca, Mg, Fe and Zn in UHT cow milks. The method was based on sorption of distinct metal species by two ion exchange columns, namely strong cation exchanger
Isabella Rustighi et al.
Journal of hazardous materials, 205-206, 10-16 (2012-01-17)
Iodinated contrast media (ICM) are persistent and ubiquitous water pollutants. Because of their high water solubility and biochemical stability, their phase-separation and recovery from the aquatic environment is very difficult. Here, borate was chosen as a complexing agent of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service