Skip to Content
Merck
All Photos(1)

Key Documents

44179

Supelco

Morphine sulfate salt pentahydrate

analytical standard

Synonym(s):

Morphine hemi[sulfate pentahydrate]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H40N2O10S · 5H2O
CAS Number:
Molecular Weight:
758.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule II; Home Office Schedule 2; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.O.OS(O)(=O)=O.CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35.CN6CC[C@]78[C@H]9Oc%10c(O)ccc(C[C@@H]6[C@@H]7C=C[C@@H]9O)c8%10

InChI

1S/2C17H19NO3.H2O4S.5H2O/c2*1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;1-5(2,3)4;;;;;/h2*2-5,10-11,13,16,19-20H,6-8H2,1H3;(H2,1,2,3,4);5*1H2/t2*10-,11+,13-,16-,17-;;;;;;/m00....../s1

InChI key

GRVOTVYEFDAHCL-RTSZDRIGSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirsten M Raehal et al.
Drug and alcohol dependence, 104(3), 187-196 (2009-06-06)
G protein-coupled receptor kinases (GRKs) are a family of intracellular proteins that desensitize and regulate the responsiveness of G protein-coupled receptors (GPCRs). In the present study, we assessed the contribution of GRK6 to the regulation and responsiveness of the G
Marsha A Raebel et al.
JAMA, 310(13), 1369-1376 (2013-10-03)
Obesity is associated with chronic noncancer pain. It is not known if opioid use for chronic pain in obese individuals undergoing bariatric surgery is reduced. To determine opioid use following bariatric surgery in patients using opioids chronically for pain control
Virginia Aguado-Lorenzo et al.
Archives of disease in childhood, 98(12), 975-979 (2013-10-24)
To investigate the accuracy of morphine infusions prepared for neonates in relation to the label strength and to identify the differences in deviation between infusions made in neonatal intensive care unit (NICU) and those dispensed ready-to-use from pharmacy. Unused portions
Serotonin, morphine, and neuropathic pain: not a simple story.
Kelly Byrne et al.
Anesthesiology, 121(2), 217-218 (2014-06-03)
Nian Gong et al.
Anesthesiology, 120(4), 962-975 (2013-08-10)
D-Amino acid oxidase (DAAO) is a flavin adenine dinucleotide-dependent peroxisomal flavoenzyme which is almost exclusively expressed within astrocytes in the spinal cord. DAAO catalyzes oxidation of D-amino acids to hydrogen peroxide, which is a stable and less active reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service