Skip to Content
Merck
All Photos(1)

Documents

30315-M

Sigma-Aldrich

Potassium iodide

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 745 °C)

grade

puriss. p.a.

Assay

≥99.0%
99.0-100.5% (calc. to the dried substance)

form

crystalline

impurities

Reducing substances, complies
≤0.0005% heavy metals (as Pb)
≤0.005% thiosulfate (S2O3)
≤0.02% free alkali (as KOH)
≤10 ppm total nitrogen (N)

loss

≤0.2% loss on drying, 105 °C

pH

6-8 (20 °C, 5%)

mp

681 °C (lit.)

anion traces

bromide, chloride (as Cl-): ≤100 ppm
iodate (IO3-): ≤2 ppm
phosphate (PO43-): ≤10 ppm
sulfate (SO42-): ≤10 ppm

cation traces

As: ≤0.00001%
Ba: ≤0.002%
Ca: ≤0.001%
Fe: ≤0.0003%
Mg: ≤0.001%
Na: ≤0.03%

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide (KI) is an inorganic potassium salt. It is used as an iodide ion source for nucleophilic displacement reactions and acts as a mediator during the electrochemical oxidations of aldehydes. In presence of an acid, KI cleaves some ethers. KI reacts with sulfonyl chlorides to produce symmetrical thiosulfonic S-esters. In the presence of oxidizing agents, KI iodinates aromatic compounds and organothallium compounds.

Application

Potassium iodide is used:
  • As a solid base catalyst loaded with alumina for transesterification of soybean oil with methanol.
  • As a iodide source in the solvothermal synthesis of BiOI (bismuth oxyiodide) microspheres using ethylene glycol solvent.
  • In the synthesis of RGO (reduced graphene oxide) from GO (graphite oxide) in presence of HCl–KI mixture.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Miyachi et al.
The British journal of dermatology, 107(2), 209-214 (1982-08-01)
The effects of potassium iodide, colchicine and dapsone on the in vitro generation of polymorphonuclear leukocyte (PMN)-derived oxygen intermediates were investigated. These three drugs have beneficial effects on those conditions in which PMNs play an important pathogenetic role. Three oxygen
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service