Skip to Content
Merck
All Photos(1)

Key Documents

12282

Millipore

Hoyle′s tellurite agar, base

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Hoyle′s Medium, Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

composition

agar, 15.0 g/L
beef extract, 10.0 g/L
peptic digest of animal tissue, 10.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.8±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

suitability

Corynebacterium spp.

Application

Hoyle Medium Base is a highly selective medium used for the isolation and differentiation of Corynebacterium diphtheriae types.

Preparation Note

Suspend 40 grams in 915 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 55°C and aseptically add 50 ml of laked blood and 35 ml of 1.0% Potassium Tellurite Solution. (Cat. No. 17774) Mix well and pour into sterile Petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry V Chistyakov et al.
Cells, 8(12) (2019-12-07)
A phenomenon of endotoxin tolerance where prior exposure of cells to minute amounts of lipopolysaccharide (LPS) causes them to become refractory to a subsequent high-amount endotoxin challenge is well described for innate immune cells such as monocytes/macrophages, but it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service